missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-8/CXCL8 (Mucin Stalk Chimera) Protein
Description
The Recombinant Human IL-8/CXCL8 (Mucin Stalk Chimera) Protein is derived from NS0. The Recombinant Human IL-8/CXCL8 (Mucin Stalk Chimera) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3576 |
| Molecular Weight (g/mol) | 34.8 kDa |
| Name | CXCL8/IL-8 Mucin Stalk Chimera |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-8/CXCL8 protein, (N-terminus) Human IL-8 (Ala23-Ser99) (Asn98Gly) & (Ser28-Ser99) (Asn98Gly) Accession &Num; P10145 Human Fractalkine Mucin-like Stalk (Phe103-Gln341) Accession &Num; P78423 KQN 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?