missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£193.30 - £343.40
Specifications
Gene ID (Entrez) | 3576 |
---|---|
Molecular Weight (g/mol) | 8.9 kDa |
Name | Endothelial Cell-Derived CXCL8/IL-8 aa 23-99 |
Source | E. coli-derived human IL-8/CXCL8 protein Ala23-Ser99 & Ser28-Ser99 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18157600
|
R&D Systems™
618-IL-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 10μg |
£193.30
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18127640
|
R&D Systems™
618-IL-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 10μg |
£193.30
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18360098
|
R&D Systems™
618-IL-050 |
£343.40
50µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18340358
|
R&D Systems™
618-IL-050/CF |
£343.40
50µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived) is derived from E. coli. The Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived) has been validated for the following applications: Bioactivity.Specifications
3576 | |
Endothelial Cell-Derived CXCL8/IL-8 aa 23-99 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
8.9 kDa | |
E. coli-derived human IL-8/CXCL8 protein Ala23-Ser99 & Ser28-Ser99 | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
Recombinant |