missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-7 Protein

Product Code. 18737046
Click to view available options
Quantity:
1 mg
10 μg
100 μg
25 μg
250 μg
Unit Size:
100µg
10µg
1mg
250µg
25µg
This item is not returnable. View return policy

Product Code. 18737046

Brand: R&D Systems™ BT007100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.100-0.500 ng/mL. The specific activity of Recombinant Human IL-7 is >1.00 x 108 units/mg, which is calibrated against the human IL-7 reference standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) M.W.-observed: 17 kDa, under reducing conditions., M.W.-theroretical: 17 kDa
Quantity 100 μg
Source E. coli-derived human IL-7 protein Asp26-His177, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.