missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-7 Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
25 μg
250 μg
Unit Size:
100µg
10µg
1mg
250µg
25µg
Description
Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.100-0.500 ng/mL. The specific activity of Recombinant Human IL-7 is >1.00 x 108 units/mg, which is calibrated against the human IL-7 reference standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.
Specifications
Specifications
| Accession Number | P13232.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3574 |
| Molecular Weight (g/mol) | M.W.-observed: 17 kDa, under reducing conditions., M.W.-theroretical: 17 kDa |
| Quantity | 10 μg |
| Source | E. coli-derived human IL-7 protein Asp26-His177, with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction