missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-6 Protein
Click to view available options
Quantity:
1 mg
200 μg
25 μg
50 μg
Unit Size:
1mg
200µg
25µg
50µg
Description
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.2-0.8 ng/mL.
Specifications
Specifications
| Accession Number | Q75MH2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and NaCl. |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | M.W. theroretical: 20.9 kDa |
| Name | IL-6 |
| Quantity | 50 μg |
| Source | E. coli-derived human IL-6 protein Pro29-Met212 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction