missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-6, Animal-Free Protein

Product Code. 30713259
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713259 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30713259 Supplier R&D Systems™ Supplier No. AFL206020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Intended for preclinical researchers who may transition to GMP IL-6 for their clinical work

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.200-0.800 ng/mL. The specific activity of Recombinant Human IL-6 is >1.00 x 108 IU/mg, which is calibrated against the human IL-6 WHO International Standard (NIBSC code: 89/548).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05231.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 3569
Molecular Weight (g/mol) M.W.-theroretical: 20.9 kDa
Quantity 20 μg
Source E. coli-derived human IL-6 protein Pro29-Met212 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbol IL6
Conjugate Unconjugated
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Purity or Quality Grade Antigen Affinity-purified
Protein IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.