missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-4 Protein

Product Code. 18786145
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18786145

Brand: R&D Systems™ BT004500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.0400-0.320 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05112.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3565
Molecular Weight (g/mol) M.W. (Observed): 13 kDa, under reducing conditions., M.W. (Theoretical): 15.1 kDa
Quantity 500 μg
Source E. coli-derived human IL-4 protein His25-Ser153, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute the 10μg size at 100μg/mL in PBS. Reconstitute all other sizes at 500μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.