missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-4 (CHO-expressed) Protein

Product Code. 18357417
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18357417 Lyophilized from a 0.2 μm filtered solution in PBS. 10µg
18176620 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18357417 Supplier R&D Systems™ Supplier No. 6507IL010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05112
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3565
Molecular Weight (g/mol) M.W. (Observed): 17-25 kDa, reducing conditions; M.W. (theoretical): 15.0 kDa
Name IL-4
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-4 protein His25-Ser153
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Monomer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbol IL4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.