missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL36RA Protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting IL-36 beta induced IL-8 secretion by human preadipocytes is less than 1.0 μg/mL, corresponding to a specific activity of >1000 IU/mg in the presence of 20 ng/mL of recombinant human IL-36 beta.
Specifications
Specifications
| Accession Number | Q9UBH0, Q9UBH0, Q9UBH0 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 26525 |
| Molecular Weight (g/mol) | 16.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of IL-36Ra/IL-1F5 as determined by LAL method. |
| Gene Alias | FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1RP3, IL36RA, IL-36RA, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?