Learn More
DiscoverX Recombinant Human IL-34
Product Code. 17322619
Encompass
Click to view available options
:
10 μg
Product Code. 17322619
Brand: DiscoverX 921305
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recombinant Human IL-34 is an agonist for CSF1R. Provided as 10μg of dry powder alongside reconstitution buffer.
Interleukin 34 (IL-34) is a secreted cytokine that binds to the colony stimulating factor 1 receptor CSF1R and stimulates monocyte proliferation; causes phosphorylation of extracellular signal-regulated kinase-1 and -2 (ERK1/2) in human monocytes; and promotes the formation of the colony-forming unit‐macrophage (CFU-M). Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 μg
Product Type Control Ligands