missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-32 alpha Protein
Description
Measured by its ability to induce TNF-alpha secretion by RAW 264.7 mouse monocyte/macrophage cellsunder serum free conditions in the presence of muramyl dipeptide (MDP). Netea, M.G. et al. (2005) Proc. Nat. Acad. Sci. 102:16309. The ED50 for this effect is 2-12 μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and DTT. |
| Gene ID (Entrez) | 9235 |
| Molecular Weight (g/mol) | 14.9 kDa |
| Name | IL-32 alpha |
| Quantity | 50 μg |
| Source | E. coli-derived human IL-32 alpha protein Cys2-Lys131 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?