Learn More
DiscoverX Recombinant Human IL-31
Product Code. 17332579
Encompass
Click to view available options
:
10 μg
Product Code. 17332579
Brand: DiscoverX 921261
Recombinant Human IL-31 is an agonist for IL31RA. Provided as 10μg of dry powder alongside reconstitution buffer.
Interleukin 31 (IL-31) is a cytokine protein and mediator of the immune response. Produced by type 2-helper T cells, it is an agonist of STAT3, STAT1, and STAT5 through its activity with the interleukin 31A receptor (IL31RA) and OSMR. May function in skin immunity and inflammation. Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 μg
Product Type Control Ligands