missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-28B/IFN-lambda 3 Protein
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.0800-0.800 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA. |
| Gene ID (Entrez) | 282617 |
| Molecular Weight (g/mol) | 20.1 kDa |
| Name | IL-28B/IFN-lambda 3 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human IL-28B/IFN-lambda 3 protein Arg30-Val200,with the variant of Lys74Arg and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?