missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-28A/IFN-lambda 2 Protein
Description
The Recombinant Human IL-28A/IFN-lambda 2 Protein is derived from NS0. The Recombinant Human IL-28A/IFN-lambda 2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 282616 |
| Molecular Weight (g/mol) | 20.8 kDa |
| Name | IL-28A/IFN-lambda 2 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-28A/IFN-lambda 2 protein, (N-terminus) Human IL-28A Val26-Val200 Accession &Num; NP_742150 DI 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?