missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-27 Ra/WSX-1/TCCR Fc Biotin Protein, CF

Product Code. 30130451
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30130451 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130451 Supplier R&D Systems™ Supplier No. BT1479050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fc Chimera, Biotinylated

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human IL-27 R alpha /WSX-1/TCCR Fc Chimera (Catalog # BT1479) binds Recombinant Human IL-27 (Catalog # 2526-IL/CF ) with an ED50 of 12.0-160 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 9466
Molecular Weight (g/mol) MolecularWeight-observed: 95-110 kDa, under reducing conditions, MolecularWeight-theroretical: 79
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human IL-27 R alpha/WSX-1/TCCR protein Human IL-27Ra (Gly34-Lys516) (N-terminus) Accession # Q6UWB1.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Conjugate Biotin
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-27R alpha/WSX-1/TCCR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.