missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-26/AK155 Monomer Protein
Description
The Recombinant Human IL-26/AK155 Monomer Protein is derived from E. coli. The Recombinant Human IL-26/AK155 Monomer Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol. |
| Gene ID (Entrez) | 55801 |
| Molecular Weight (g/mol) | 17.7 kDa |
| Name | IL-26/AK155 Monomer |
| Quantity | 25 μg |
| Source | E. coli-derived human IL-26/AK155 protein Lys22-Gln171,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Structural Form | Monomer and a small amount of disulfide-linked homodimer (no more than 7%) |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?