missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-26/AK155 Dimer Protein

Product Code. 18118070
Click to view available options
Formulation:
Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18118070

Brand: R&D Systems™ 1870IL010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-26/AK155 Dimer Protein is derived from E. coli. The Recombinant Human IL-26/AK155 Dimer Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol with BSA as a carrier protein.
Gene ID (Entrez) 55801
Molecular Weight (g/mol) 17.7 kDa (monomer)
Name IL-26/AK155 Dimer
Quantity 10 μg
Source E. coli-derived human IL-26/AK155 protein Lys22-Gln171,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Structural Form Disulfide-linked homodimer and a small amount of monomer (no more than 10%)
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.