missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 24 Protein
A cDNA sequence encoding the IL 24 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3875.00
Specifications
Gene ID (Entrez) | 11009 |
---|---|
Name | IL 24 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | IL24 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964918
|
enQuireBio™
QP10705-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944918
|
enQuireBio™
QP10705-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954918
|
enQuireBio™
QP10705-1MG |
1 mg |
£3875.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
11009 | |
Research Use Only | |
IL24 | |
Recombinant Protein | |
S. cerevisiae | |
MNFQQRLQSL WTLARPFCPP LLATASQMQM VVLPCLGFTL LLWSQVSGAQ GQEFHFGPCQ VKGVVPQKLW EAFWAVKDTM QAQDNITSAR LLQQEVLQNV SDAESCYLVH TLLEFYLKTV FKNYHNRTVE VRTLKSFSTL ANNFVLIVSQ LQPSQENEMF SIRDSAHRRF LLFRRAFKQL DVEAALTKAL GEVDILLTWM QKFYKL | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 24 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to bind to the cell receptor of Capan-1 cells line resulted in Stat-3 activation. The ED50 for this effect is typically 1.0 ng/ml, corresponding to a Specific Activity of 1x106 units/mg. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in PBS with BSA as a carrier. |