missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-23 (CHO Expressed) Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
Description
Measured by its ability to induce IL-17 secretion by mouse splenocytes. Aggarwal, S. et al. (2003) J. Biol. Chem. 278:1910. The ED50 for this effect is 0.100-1.00 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 51561 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 59-68 kDa, 46-52 kDa & 28-33 kDa, under reducing conditions., MolecularWeight-theroretical: 55 kDa (IL-23 single chain), 37 kDa (p40) & 19 kDa (p19) |
| Quantity | 1 mg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IL-23 protein Human IL-23 p40 (Ile23-Ser328) Accession # P29460.1 GSGSSRGGSGSGGSGGGGSKL Human IL-23 p19 (Arg20-Pro189) Accession # AAG37232.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | IL23, IL-23, IL-23 p19/IL-12 p40, IL-23 A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction