missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL2 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.1 ng/mL, corresponding to a specific activity of >1.0 x 107 IU/mg.
Specifications
Specifications
| Accession Number | P60568, P60568, P60568 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 3.5. |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | 15.4 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1.0 EU/ug of IL-2 as determined by LAL method. |
| Gene Alias | aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?