Learn More
DiscoverX Recombinant Human IL-2, Cell Culture Grade
Product Code. 17362639
Encompass
Click to view available options
:
10 μg
Product Code. 17362639
Brand: DiscoverX 921331
Recombinant Human IL-2, Cell Culture Grade is an agonist for IL2RA and IL2RB. Provided as 10μg of dry powder alongside reconstitution buffer.
Interleukin 2 (IL-2) is a cytokine protein and mediator of the immune response. An agonist for IL2RA and IL2RB, IL-2 regulates activation of T and B lymphocyte proliferation, and also stimulates monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells. This cell culture grade recombinant human IL-2 is a variant form of IL-2 (contains a serine to cysteine substitution) that can be used for differentiation studies, functional assays, and cell culture. Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 μg
Product Type Control Ligands