missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-1ra/IL-1F3/IL1RN Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
500µg
Description
IL-1ra/IL-1F3/IL1RN protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting IL-1alpha-dependent proliferation of murine D10S cells is less than 40 ng/mL, corresponding to a specific activity of > 2.5 x 10^4 IU/mg in the presence of 50 pg/mL rHuIL-1alpha
Specifications
Specifications
| Accession Number | P18510, P18510, P18510 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3557 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 17.1 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of IL-1 ra/IL-1F3/IL1RN as determined by LAL method. |
| Gene Alias | DIRA, ICIL-1 RA, IL1F3, IL-1 ra, IL1RA, IL-1 ra3, IL1RN, IL-1 RN, interleukin 1 receptor antagonist, IRAP, MVCD4 |
| Gene Symbol | IL1RN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction