missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-18 R alpha/IL-1 R5 Avi His Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-18 R beta/IL-1 R7 Fc (Catalog # 118-AP ) is immobilized at 0.500 μg/mL (100 μL/well), in the present of Recombinant Human IL-18 (Catalog # 9124-IL ) Biotinylated Recombinant Human IL-18 R alpha/IL-1 R5 Avi-tag His-tag (Catalog # AVI11179) binds with an ED50 of 20.0-200 ng/mL. The biotin to protein ratio is great than 0.7 as determined by the HABA assay.
Specifications
Specifications
| Accession Number | Q13478.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 63-71 kDa, under reducing conditions., M.W.-theroretical: 40 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IL-18 R alpha/IL-1 R5 protein Human IL-18 R alpha/IL-1 R5 (Cys22-Arg329) (C-terminus) Accession # Q13478.1 Avi-tag 6-His tag (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Biotin |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction