missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-18/IL-1F4 Protein, Animal-Free

Product Code. 30689938
Change view
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689938 50 μg 50µg
30689919 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30689938 Supplier R&D Systems™ Supplier No. BT018AFL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED50 for this effect is 1.25-15.0 ng/mL. The specific activity of GMP Recombinant Human IL-18/IL-1F4 is >5.00 x 106 units/mg, which is calibrated against the human IL-18 WHO Standard (NIBSC code: 03/200).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14116.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and TCEP with Trehalose.
Gene ID (Entrez) 3606
Molecular Weight (g/mol) M.W.-observed: 18 kDa, under reducing conditions., M.W.-theroretical: 18 kDa
Quantity 50 μg
Source E. coli-derived human IL-18/IL-1F4 protein Tyr37-Asp193 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in water.
Protein IL-18/IL-1F4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.