missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-18/IL-1F4 Protein

Product Code. 18715147
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Unit Size:
10µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18715147

Brand: R&D Systems™ 9124IL500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Human IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14116
For Use With (Application) Bioactivity
Gene ID (Entrez) 3606
Molecular Weight (g/mol) Predicted MW: 18kDa Observed MW: 18kDa, reducing conditions
Quantity 500 μg
Source E. coli-derived human IL-18/IL-1F4 protein Tyr37-Asp193
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Common Name IL-18/IL-1F4
Gene Symbol IL18
Biological Activity Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED50 for this effect is typically 1.5-9ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Tyr37-Asp193
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.