Learn More
DiscoverX Recombinant Human IL-17F
Product Code. 17312579
Encompass
Click to view available options
:
25 μg
Product Code. 17312579
Brand: DiscoverX 921259
Recombinant Human IL-17F is an agonist for IL17RA. Provided as 25μg of dry powder alongside reconstitution buffer.
Interleukin 17F (IL-17F) is a cytokine protein and one of the variants of IL-17. An agonist for IL17RA, IL-17F stimulates production of IL-6, IL-8, and granulocyte colony-stimulating factor cytokines, as well as PBMC and T-cells. Inhibits angiogenesis, and can regulate cartilage matrix turnover. Provided as 25μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 25 μg
Product Type Control Ligands