missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-17B protein is fully biologically active when compared to standard. The ED50 as determined by inducing IL-8 secretion of human HepG2 cells is less than 1.0 μg/mL, corresponding to a specific activity of >1000 IU/mg
Specifications
Specifications
| Accession Number | Q9UHF5, Q9UHF5, Q9UHF5 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, with 0.1 % Tween-20 and 3 % Trehalose. |
| Gene ID (Entrez) | 27190 |
| Molecular Weight (g/mol) | 36.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of IL-17B as determined by LAL method. |
| Gene Alias | Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?