missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-17AF Protein

Recombinant Protein

Brand:  enQuireBio™ QP5200-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955649

  • £715.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002181.1
IL-17AF Protein
Research Use Only
IL17
Recombinant Protein
E. coli
IL-17A:MIV KAGITIPRNP GCPNSEDKNF PRTVMVNLNI HNRNTNTNPK RSSDYYNRST SPWNLHRNED PERYPSVIWE AKCRHLGCIN ADGNVDYHMN SVPIQQEILV LRREPPHCPN SFRLEKILVS VGCTCVTPIV HHVA IL-17F :MRKIPKVGH TFFQKPESCP PVPGGSMKLD IGIINENQRV SMSRNIESRS TSPWNYTVTW DPNRYPSEVV QAQCRNLGCI NAQGKEDISM NSVPIQQETL VVRRKHQGCS VSFQLEKVLV TVGCTCVTPV IHHVQ
Greater than 95% as determined by SDS-PAGE.
3605
100 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Production of IL-6 from mouse 3T3 cells was significantly induced by less than 50 ng/ml of Human IL-17AF, or activity of at least 2.0 x 10^4 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers