missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-17A/F Heterodimer Biotin Protein

Product Code. 18772908
Click to view available options
:
25μg; Biotin
This item is not returnable. View return policy

Product Code. 18772908

Brand: R&D Systems™ BT5837025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-17A/F Heterodimer Biotin Protein is derived from CHO. The Recombinant Human IL-17A/F Heterodimer Biotin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3605
Molecular Weight (g/mol) Theoretical M.W.: 15kDa (unlabeled IL-17A), 15kDa (unlabeled IL-17F)
Quantity 25μg
Source Chinese Hamster Ovary cell line,CHO-derived human IL-17A/F Heterodimer protein, (N-terminus) Human IL-17A (Gly24-Ala155) Accession &Num; Q16552 Human IL-17F (Arg21-GLN163) Accession &Num; Q96PD4 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias CTLA8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, Cytotoxic T-Lymphocyte-Associated Protein 8, IL17, IL-17, IL-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A, interleukin-17A
Common Name IL-17A/F Heterodimer
Biological Activity Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.4-2ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human IL-17A (Gly24-Ala155) Accession # Q16552, N-terminus; Human IL-17F (Arg21-GLN163) Accession # Q96PD4, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.