missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.300-2.60 ng/mL. The specific activity of recombinant human IL-15 is greater or equal to 1.50 x 10^8 units/mg
Specifications
Specifications
| Accession Number | P40933, P40933, P40933 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3600 |
| Molecular Weight (g/mol) | 12.9 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of IL-15 as determined by LAL method. |
| Gene Alias | IL-15MGC9721, interleukin 15, interleukin-15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?