missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-15 Protein
A DNA sequence encoding the Homo sapiens (Human) IL-15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8555-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000576.1 | |
IL-15 Protein | |
Research Use Only | |
IL15 | |
Recombinant Protein | |
E. coli | |
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SLESGDASIH DTVENLIILA NNSLSSNGNV TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS | |
Greater than 90% as determined by SDS-PAGE. |
3600 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |