missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-15 Protein
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
Description
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.3-2.6 ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554). Specific activity is for reference purposes only and is not routinely tested.
Specifications
Specifications
| Accession Number | P40933.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Reconstitute at 100μg/mL in sterile PBS. |
| Gene ID (Entrez) | 3600 |
| Molecular Weight (g/mol) | 10 kDa, under reducing conditions. |
| Quantity | 5 μg |
| Source | E. coli-derived human IL-15 protein Asn49-Ser162 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | IL15, IL-15MGC9721, interleukin 15, interleukin-15 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction