missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-13 R alpha 1 Fc Avi-tag Protein
Description
Measured by its binding ability in a functional ELISA. When Biotinylated Recombinant Human IL-13 R alpha 1 Fc Chimera Avi-tag (Catalog # AVI10436) is immobilized at 1 μg/mL (100 μL/well), Recombinant Human IL-13 (Catalog # 213-ILB ) binds with an ED50 of 10-80 ng/mL.
Specifications
Specifications
| Accession Number | AAB37127.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3597 |
| Molecular Weight (g/mol) | M.W.-theroretical: 65 kDa, M.W.-observed: 93-103 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IL-13 R alpha 1 protein Human IL-13 R alpha 1 (Ala27-Thr343) (c-terminus) Accession # AAB37127.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL13R alpha 1, IL-13 R subunit alpha-1, IL-13 Ra, IL13RA, IL13RA1, IL-13 RA1, IL-13 R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?