missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 12 p35 Protein
A cDNA sequence encoding the IL 12 p35 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12391-10ug
Additional Details : Weight : 0.01000kg
Specifications
3592 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MCPARSLLLV ATLVLLDHLS LARNLPVATP DPGMFPCLHH SQNLLRAVSN MLQKARQTLE FYPCTSEEID HEDITKDKTS TVEACLPLEL TKNESCLNSR ETSFITNGSC LASRKTSFMM ALCLSSIYED LKMYQVEFKT MNAKLLMDPK RQIFLDQNML AVIDELMQAL NFNSETVPQK SSLEEPDFYK TKIKLCILLH AFRIRAVTID RVMSYLNAS | |
Greater than 95.0% as determined by SDS-PAGE. |
IL 12 p35 Protein | |
Research Use Only | |
IL12A | |
Human | |
His | |
Interleukin-12 p35 His-Tag is supplied in 1xPBS and 50% glycerol. |