missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-12 (linked heterodimer) Protein

Product Code. 18156890
Change view
Click to view available options
Quantity:
10 μg
20 μg
50 μg
Unit Size:
10µg
20µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18156890 10 μg 10µg
18709626 50 μg 50µg
18360568 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18156890 Supplier R&D Systems™ Supplier No. 10018IL010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-12 (linked heterodimer) Protein is derived from HEK293. The Recombinant Human IL-12 (linked heterodimer) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29460.1 (IL-12p40), P29459.2 (IL-12p35)
For Use With (Application) Bioactivity
Gene ID (Entrez) 3592
Molecular Weight (g/mol) Observed M.W.: 63-75 kDa, under reducing conditions.
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human IL-12 protein Human IL-12p40 (Ile23-Ser328) (N-terminus) Accession # P29460.1 GSGSSRGGSGSGGSGGGGSK Human IL-12p35 (Arg23-Ser219) (N-terminus) Accession # P29459.2(C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity IL-12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100-200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.