missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-12/IL-23 p40 Protein, CF

Product Code. 18706105
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
Unit Size:
100µg
10µg
1mg
250µg
50µg
This item is not returnable. View return policy

Product Code. 18706105

Brand: R&D Systems™ 11407IL01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(HEK293-Expressed)

Measured by its ability to enhance IFN-gamma secretion in NK-92 human natural killer lymphoma cells. The ED50 for this effect is 2.00-20.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29460.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3593
Molecular Weight (g/mol) M.W. (Observed): 38-45 kDa, under reducing conditions., M.W. (Theroretical): 35 kDa
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived human IL-12/IL-23 p40 protein Ile23-Ser328
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL and other sizes at 500 μg/mLin PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-12/IL-23 p40
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.