missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-11 R alpha Protein

Product Code. 18765677
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18765677

Brand: R&D Systems™ 8895MR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-11 R alpha Protein is derived from CHO. The Recombinant Human IL-11 R alpha Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14626
For Use With (Application) Bioactivity
Gene ID (Entrez) 3590
Molecular Weight (g/mol) Observed M.W.: 40-55kDa, reducing conditions, Theoretical M.W.: 38kDa
Name IL-11R alpha
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human IL-11 R alpha protein Ser24-Val363,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1
Common Name IL-11 R alpha
Gene Symbol IL11RA
Biological Activity Measured by its ability to bind recombinant human IL-11 in a functional ELISA with an estimated Kd <1 nM.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser24-Val363, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.