missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-10 R beta Fc Chimera Protein, CF
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human IL-10 R beta Fc Chimera (Catalog # 11460-RB) binds Recombinant Human IL-28 R alpha/IFN-lambda R1 Fc Chimera (Catalog # 5260-MR ) in the presence of Recombinant Human IL-28B/IFN-lambda 3 (Catalog # 5259-IL/CF ) with an ED50 of 0.250-3.00 μg/mL.
Specifications
Specifications
| Accession Number | Q08334.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3588 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 72-82 kDa, under reducing conditions., MolecularWeight-theroretical: 50 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IL-10 R beta protein Human IL-10 R beta (Met20-Ser220) (N-terminus) Accession # Q08334.2 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction