missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-10 Protein
Description
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.075-0.750 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3586 |
| Molecular Weight (g/mol) | 18.6 kDa |
| Name | IL-10 |
| Quantity | 5 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-10 protein Ser19-Asn178 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homodimer |
| Gene Alias | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?