missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-1 RI Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-1 RI Fc Chimera (Catalog # 11085-RI) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Human IL-1 beta/IL-1F2 (Catalog # 201-LB ) binds with an ED50 of 0.600-5.40 ng/mL.
Specifications
Specifications
| Accession Number | P14778.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3554 |
| Molecular Weight (g/mol) | M.W.-observed: 80-90 kDa, under reducing conditions., M.W.-theroretical: 63 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IL-1 RI protein Human IL-1 RI (Asp21-Lys336) (C-terminus) Accession # P14778.1 IEGRMD Human IgG1 Fc (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1 RI, IL1RT1, IL-1 RT1, IL-1 RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?