missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1 alpha / IL1A / IL1F1 Protein
Specifications
Specifications
| Accession Number | NP_000566.3 |
| Gene ID (Entrez) | 3552 |
| Name | IL-1 alpha / IL1A / IL1F1 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | IL1A |
| Biological Activity | To pass QC, D10S cell proliferation must be significantly induced by less than 50 pg/ml or human IL-1 alpha / IL1A / IL1F1 (or activity must be at least 2.0 x 10^7 units/mg). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?