missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IHH Protein

A DNA sequence encoding the Homo sapiens (Human) IHH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7597-ec-1mg

 View more versions of this product

Product Code. 16049650

  • £1735.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_002172.2
IHH Protein
Research Use Only
IHH
Recombinant Protein
E. coli
CFPAGAQVRL ESGARVALSA VRPGDRVLAM GEDGSPTFSD VLIFLDREPH RLRAFQVIET QDPPRRLALT PAHLLFTADN HTEPAARFRA TFASHVQPGQ YVLVAGVPGL QPARVAAVST HVALGAYAPL TKHGTLVVED VVASCFAAVA DHHLAQLAFW PLRLFHSLAW GSWTPGEGVH WYPQLLYRLG RLLLEEGSFH PLGMSGAGS
Greater than 90% as determined by SDS-PAGE.
3549
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers