missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from 0.2um-filtered solution in PBS |
| Molecular Weight (g/mol) | M.W. Theoretical: 32 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <5EU/mg purified protein (LAL test; Lonza). |
| Reconstitution | Reconstitute with 100 ul sterile water. Add 1X PBS to the desired protein concentration |
| Research Category | Immunology |
| Purity or Quality Grade | >98%, by SDS-PAGE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?