missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IgG1 Fc Protein
Description
The Recombinant Human IgG1 Fc Protein is derived from NS0. The Recombinant Human IgG1 Fc Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Source | Mouse myeloma cell line,NS0-derived human IgG1 protein, (N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) Substitutions: Leu117Ala,Leu118Glu,Gly120Ala,Thr276Ala Accession &Num; P01857 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Dimer |
| Gene Alias | IgG1, Igh-4, RGD1359539, VH7183 |
| Conjugate | Unconjugated |
| Reconstitution | It is recommended that sterile PBS containing at least 0.1% human serum albumin or bovine serum albumin be added to the vial to prepare a stock solution of no less than 100 μg/mL. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?