missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IgG receptor FcRn large subunit p51 Protein
A DNA sequence encoding the Homo sapiens (Human) IgG receptor FcRn large subunit p51, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6030-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129491.1 | |
IgG receptor FcRn large subunit p51 Protein | |
Research Use Only | |
FCGRT | |
Recombinant Protein | |
Yeast | |
AESHLSLLYH LTAVSSPAPG TPAFWVSGWL GPQQYLSYNS LRGEAEPCGA WVWENQVSWY WEKETTDLRI KEKLFLEAFK ALGGKGPYTL QGLLGCELGP DNTSVPTAKF ALNGEEFMNF DLKQGTWGGD WPEALAISQR WQQQDKAANK ELTFLLFSCP HRLREHLERG RGNLEWKEPP SMRLKARPSS PGFSVLTCSA FSFYPPELQL RFLRNGLAAG TGQGDFGPNS DGSFHASSSL TVKSGDEHHY CCIVQHAGLA QPLRVELESP AKSS | |
Greater than 90% as determined by SDS-PAGE. |
2217 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |