missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IGFBP-rp1/IGFBP-7 Protein

Product Code. 18407111 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407111 100 μg 100µg
18406421 5 μg 5µg
18429291 500 μg 500µg
18459601 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18407111 Supplier Novus Biologicals™ Supplier No. NBP235007100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16270, Q16270, Q16270
Concentration LYOPH
For Use With (Application) PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 8.6, 150 mM NaCl.
Gene ID (Entrez) 3490
Molecular Weight (g/mol) 26.4 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/ug of IGFBP-rp1/IGFBP-7 as determined by LAL method.
Gene Alias angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor
Gene Symbol IGFBP7
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein IGFBP-rp1/IGFBP-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.