missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fully biologically active when compared to standard. The ED50 as determined by inhibiting IGF-II induced proliferation of serum free human MCF-7 cells is less than 200 ng/mL, corresponding to a specific activity of >5.0 x 10^3 IU/mg in the presence of 15 ng/mL of rHuIGF-II.
Specifications
Specifications
| Accession Number | P17936, P17936, P17936 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3486 |
| Molecular Weight (g/mol) | 28.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of IGFBP-3 as determined by LAL method. |
| Gene Alias | acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?