missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGF1 Protein

A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12380-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960229

  • £302.00 / 5µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3479
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
IGF1 is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
IGF1 Protein
Research Use Only
IGF1
Human
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers