missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-lambda 4 Protein

Product Code. 18798406
Click to view available options
:
Recombinant Human IFN-lambda 4 Protein, Carrier-Free
This item is not returnable. View return policy

Product Code. 18798406

Brand: R&D Systems™ 9165IF025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IFN-lambda 4 Protein is derived from E. coli. The Recombinant Human IFN-lambda 4 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number K9M1U5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Citric Acid and CHAPS.
Gene ID (Entrez) 101180976
Molecular Weight (g/mol) 18 kDa
Quantity 25μg
Source E. coli-derived human IFN-lambda 4 protein Ala23-Leu179 with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Gene Alias IFNAN, IFNL4, IFNlambda 4
Biological Activity Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.2-1.2 μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.