missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-alpha 21/IFNA21 Protein

Product Code. 18739746
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18739746

Brand: R&D Systems™ 11159IF010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 1.00-30.0 pg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01568.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3452
Molecular Weight (g/mol) M.W.-observed: 18-22 kDa, under reducing conditions., M.W.-theroretical: 19 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IFN-alpha 21/IFNA21 protein Cys24-Glu189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IFNA21, IFNalpha F, IFN-alpha F, IFN-alpha-21, IFN-alphaI, interferon alpha-21, Interferon alpha-F, interferon, alpha 21, LeIF F, leukocyte interferon protein, MGC126687, MGC126689
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IFN-alpha F/IFNA21
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.